Product Name
β-Ionone natural, ≥85%, FG
Product Number
W259519
Product Brand
ALDRICH
CAS Number
14901-07-6
Molecular Weight
192.30
Test
Specification
Appearance (Color)Colorless to Yellow
Appearance (Form)Liquid
Refractive index at 20 ° C1.515 - 1.525
Infrared spectrumConforms to Structure
Purity (GC)≥85.00 %
Specific Gravity at 25 °C0.940 - 0.950 g/cm3
13Carbon Analysis≥1.00 ppm
14Carbon Analysis≥1.00 dpm/gC
Deuterium Analysis≥1.0 ppm
 The above analysis is consistent with that found
 for material of natural origin. This information
 is non-transferrable; it is for the sole and
 exclusive use of the immediate buyer.
Arsenic (As)≤3 ppm
Cadmium (Cd)≤1 ppm
Mercury (Hg)≤1 ppm
Lead (Pb)≤10 ppm