Product Name
Skatole ≥98%
Product Number
W301906
Product Brand
ALDRICH
CAS Number
83-34-1
Molecular Weight
131.17
Test
Specification
Appearance (Color)White to Faint Yellow
Appearance (Form)Solid
Melting Point92.0 - 97.0 °C
Infrared spectrumConforms to Structure
Purity (GC)≥98.0 %
Heavy Metal≤10 ppm
Arsenic (As)≤3 ppm
Cadmium (Cd)≤1 ppm
Mercury (Hg)≤1 ppm
Lead (Pb)≤1 ppm