Product Name
p-Anisic acid ≥99%
Product Number
W505404
Product Brand
ALDRICH
CAS Number
100-09-4
Molecular Weight
152.15
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
MELTING POINT183 - 186 DEGREES CELSIUS
INFRARED SPECTRUMCONFORMS TO STRUCTURE.
TITRATION99.0% - 101.0% (WITH NAOH)
THIN-LAYERCONSISTENT WITH 99+% PURITY
SOLUBILITY5% IN 95% ETHANOL; CLEAR, COLORLESS TO PALE
 YELLOW SOLUTION
 REVISED APRIL 5, 2001 JSB