Product Name
N-Succinylglycine analytical standard
Product Number
01883
Product Brand
SIAL
CAS Number
5694-33-7
Molecular Weight
175.14
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS