Product Name
2,6-Pyridinedicarboxylic acid suitable for ion chromatography, ≥99.5% (T)
Product Number
02321
Product Brand
SIAL
CAS Number
499-83-2
Molecular Weight
167.12
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M≥ 99.5 %
PURITY (GC AREA %)≥ 99.5 %
RESIDUE ON IGNITION≤ 0.05 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
ALUMINIUM (ICP)≤ 10 mg/kg
BARIUM (ICP)≤ 10 mg/kg
BISMUTH (ICP)≤ 10 mg/kg
CALCIUM (ICP)≤ 10 mg/kg
CADMIUM (ICP)≤ 5.0 mg/kg
COBALT (ICP)≤ 5.0 mg/kg
CHROMIUM (ICP)≤ 5.0 mg/kg
COPPER (ICP)≤ 5.0 mg/kg
IRON (ICP)≤ 2.0 mg/kg
POTASSIUM (ICP)≤ 100 mg/kg
LITHIUM (ICP)≤ 5.0 mg/kg
MAGNESIUM (ICP)≤ 5.0 mg/kg
MANGANESE (ICP)≤ 5.0 mg/kg
MOLYBDENUM (ICP)≤ 5.0 mg/kg
SODIUM (ICP)≤ 50 mg/kg
NICKEL (ICP)≤ 5.0 mg/kg
LEAD (ICP)≤ 5.0 mg/kg
STRONTIUM (ICP)≤ 5.0 mg/kg
ZINC (ICP)≤ 5.0 mg/kg
PHOSPHATE (PO4)≤ 5.0 mg/kg
RECOMMENDED RETEST PERIOD42 MONTHS