Product Name
Ethylbenzene analytical standard
Product Number
03079
Product Brand
SIAL
CAS Number
100-41-4
Molecular Weight
106.17
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.5 %
REMARKS ON GC--
REFRACTIVE INDEX N20/D1.494 - 1.496
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS