Product Name
N-[2-(4-Hydroxyphenyl)acetyl]glycine analytical standard
Product Number
03417
Product Brand
SIAL
CAS Number
28116-23-6
Molecular Weight
209.20
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS