Product Name
N-Pivaloylglycine analytical standard
Product Number
03535
Product Brand
SIAL
CAS Number
23891-96-5
Molecular Weight
159.18
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS