Product Name
γ-Aminobutyric acid analytical standard
Product Number
03835
Product Brand
SIAL
CAS Number
56-12-2
Molecular Weight
103.12
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 97.0 %
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS