Product Name
Scintillation cocktail for radiometry
Product Number
03999
Product Brand
SIAL
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)COLORLESS TO VERY FAINTLY BLUE OR VERY FAINTLY VIOLET
APPEARANCE (FORM)Liquid or Viscous Liquid
DENSITY D20/40.982 - 0.987
REFRACTIVE INDEX N20/D1.527 - 1.531