Product Name
Valeryl-L-carnitine analytical standard
Product Number
04265
Product Brand
SIAL
CAS Number
40225-14-7
Molecular Weight
245.32
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
SPECIFIC ROTATION (20/D)-20.0 ± 2.0 Degrees
CONCENTRATIONC=0.1 in Water
ELEMENT ANALYSIS (CALC.)≤ 10 % water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS