Product Name
3-Hexanol analytical standard
Product Number
04649
Product Brand
SIAL
CAS Number
623-37-0
Molecular Weight
102.17
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
REFRACTIVE INDEX N20/D1.415 - 1.417
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS