Product Name
3-(2-Aminoethylamino)propyldimethoxymethylsilane ≥95.0%
Product Number
06667
Product Brand
SIAL
CAS Number
3069-29-2
Molecular Weight
206.36
Test
Specification
APPEARANCE (COLOR)Colorless to Very Light Yellow and Colorless to Very Light Green-Yellow
APPEARANCE (FORM)Liquid
TITRATION (NT) HCLO4 0.1M95.0 - 105.0 %
REFRACTIVE INDEX N20/D1.448 - 1.452
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE