Product Name
3,4-Dihydroxyphenylacetic acid analytical standard
Product Number
11569
Product Brand
SIAL
CAS Number
102-32-9
Molecular Weight
168.15
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M97.0 - 103.0 %
PURITY (HPLC AREA %)≥ 97.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS