Product Name
2,2′-Azobis(2-methylpropionitrile) purum, ≥98.0% (GC)
Product Number
11630
Product Brand
SIAL
CAS Number
78-67-1
Molecular Weight
164.21
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals
PURITY (GC AREA %)≥ 98.0 %
SOLUBILITY (COLOR)Colorless
SOLUBILITY (TURBIDITY)Clear
SOLUBILITY (METHOD)0.5G IN 10ML TOLUENE
WATER≤ 2.0 %
RESIDUE ON IGNITION≤ 0.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE