Product Name
Capsaicin analytical standard
Product Number
12084
Product Brand
SIAL
CAS Number
404-86-4
Molecular Weight
305.41
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.5 %
MELTING POINT66 - 69 C
CARBON CONTENT70.79 % (THEORY)
HYDROGEN CONTENT8.91 % (THEORY)
NITROGEN CONTENT4.59 % (THEORY)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD48 MONTHS