Product Name
Benzene analytical standard
Product Number
12540
Product Brand
SIAL
CAS Number
71-43-2
Molecular Weight
78.11
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.9 %
REFRACTIVE INDEX N20/D1.500 - 1.502
WATER (COULOMETR.)≤ 0.03 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS