Product Name
Benzyl cyanide analytical standard
Product Number
12608
Product Brand
SIAL
CAS Number
140-29-4
Molecular Weight
117.15
Test
Specification
APPEARANCE (COLOR)Colorless to Light Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.520 - 1.526
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS