Product Name
Potassium (poly)sulfide ≥42% K2S basis
Product Number
12665
Product Brand
SIAL
CAS Number
37199-66-9
Molecular Weight
Test
Specification
REDOX TITRATION≥ 42 % as K2S
REDOXTITRATION (METHOD)IODOMETRIC
RECOMMENDED RETEST PERIOD48 Months