Product Name
1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid ≥96.0% (HPLC)
Product Number
14510
Product Brand
SIAL
CAS Number
85233-19-8
Molecular Weight
476.43
Test
Specification
APPEARANCE (COLOR)White to Faint Beige
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 96.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE