Product Name
Serotonin analytical standard
Product Number
14927
Product Brand
SIAL
CAS Number
50-67-9
Molecular Weight
176.22
Test
Specification
APPEARANCE (COLOR)White to Dark Grey and Faint Brown to Dark Brown
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
CARBON CONTENT68.16% (Theory)
HYDROGEN CONTENT6.86% (Theory)
NITROGEN CONTENT15.90% (Theory)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS