Product Name
Brilliant Cresyl blue solution for microscopy, 0.3 % (w/v) in ethanol
Product Number
16035
Product Brand
SIAL
CAS Number
51716-96-2
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Blue to Very Dark Blue
APPEARANCE (FORM)Liquid
THIN LAYER CHROMATOGR.CORRESPONDS TO STANDARD
DENSITY D20/40.798 - 0.818
REFRACTIVE INDEX N20/D1.360 - 1.370
EXTINCTION (1%/1CM) (1) (UV)--
SOLVENT (UV)ETOH 50 %
EXPIRY PERIOD30 MONTHS