Product Name
Hydrogen chloride – 2-propanol solution ~1.25 M HCl (T), for GC derivatization, LiChropur
Product Number
17933
Product Brand
SIAL
CAS Number
Molecular Weight
36.46
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
TITRATION (MOLARITY)1.12 - 1.38 mol/l
PURITY (GC AREA %)≥ 99.0 %
DENSITY D20/40.816 - 0.826
REFRACTIVE INDEX N20/D1.340 - 1.440
WATER≤ 0.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD42 MONTHS