Product Name
1-Methylpyrrole analytical standard
Product Number
18698
Product Brand
SIAL
CAS Number
96-54-8
Molecular Weight
81.12
Test
Specification
APPEARANCE (COLOR)Colorless to Brown-Yellow and Colorless to Brown and Colorless to Orange
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.0 %
REFRACTIVE INDEX N20/D1.488 - 1.490
WATER (COULOMETR.)≤ 0.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS