Product Name
2-Hexanone purum, ≥96.0% (GC)
Product Number
20270
Product Brand
SIAL
CAS Number
591-78-6
Molecular Weight
100.16
Test
Specification
APPEARANCE (COLOR)Colorless to Very Faint Yellow and Colorless to Very Faint Green-Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 96.0 %
REFRACTIVE INDEX N20/D1.400 - 1.402
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE