Product Name
(−)-Camphor-10-sulfonic acid purum, ≥98.0% (T)
Product Number
21365
Product Brand
SIAL
CAS Number
35963-20-3
Molecular Weight
232.30
Test
Specification
APPEARANCE (COLOR)White to Faint Brown and Faint Beige
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 1M98.0 - 102.0 %
THIN LAYER CHROMATOGR.CORRESPONDS TO STANDARD
SPECIFIC ROTATION (20/D)-21.5 ± 1.0 Degrees
CONCENTRATIONC=10 IN WATER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD54 MONTHS