Product Name
Disperse Blue 1 Dye content 30 %
Product Number
215643
Product Brand
SIAL
CAS Number
2475-45-8
Molecular Weight
268.27
Test
Specification
APPEARANCEGREY TO BLACK POWDER
INFRARED SPECTRUMCONFORMS TO STRUCTURE
UV-VISIBLE SPECTRUM0.01G/L IN 50% ETHANOL W/1 DROP 1N NAOH
 E(610 +/- 15NM)>5,000
 REVISED JUNE 13, 2000 JSB