Product Name
Silica gel high-purity grade, pore size 60 Å, 2-25 μm particle size, without binder, pore volume 0.75 cm3/g, suitable for thin layer chromatography (TLC)
Product Number
288500
Product Brand
SIAL
CAS Number
112926-00-8
Molecular Weight
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder
VENDOR INFORMATIONSURFACE AREA: 400 +/-50 M2/G, N2*
PORE VOLUME: 0.75 +/- 0.1 ML/G, N2*
PARTICLE SIZE, MEDIAN: 12 +/-4 UM*
PARTICLE SIZE, DISTRIBUTION: D95/D5 <7%*
PH: 5.5-7.5*
LOSS ON DRYING: <9%*
REMARKS* SUPPLIER SPECIFICATIONS