Product Name
3-Methylcrotonyl-L-carnitine analytical standard
Product Number
28986
Product Brand
SIAL
CAS Number
64656-41-3
Molecular Weight
243.30
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 95.0 %
SPECIFIC ROTATION (20/D)-18 ± 2 Degrees
CONCENTRATIONC=0.1 in Water
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS