Product Name
Decane analytical standard
Product Number
30540
Product Brand
SIAL
CAS Number
124-18-5
Molecular Weight
142.28
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 99.8 %
REFRACTIVE INDEX N20/D1.411 - 1.413
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS