Product Name
Sodium 1-decanesulfonate suitable for ion pair chromatography, LiChropur, ≥99.0%
Product Number
30631
Product Brand
SIAL
CAS Number
13419-61-9
Molecular Weight
244.33
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals or Flakes
TITRATION (ION EXCHANGE)≥ 99.0 %
SUITABILITY TEST (HPLC)CORRESPONDS TO STANDARD
CARBON CONTENT49.16 % (THEORY)
HYDROGEN CONTENT8.66 % (THEORY)
INFRARED SPECTRUMCONFORMS TO STRUCTURE
UV Absorbance 210nm≤ 0.05
UV Absorbance 220nm≤ 0.03
UV Absorbance 230nm≤ 0.02
UV Absorbance 260nm≤ 0.02
UV Absorbance 500nm≤ 0.02
CYCLIC VOLTAMMOGRAMCORRESPONDS TO STANDARD CV
RESIDUE (FILTER TEST)NO RESIDUE
FILTERTEST (METHOD)5% in Water
RECOMMENDED RETEST PERIOD42 MONTHS