Product Name
Prosulfocarb PESTANAL®, analytical standard
Product Number
31141
Product Brand
SIAL
CAS Number
52888-80-9
Molecular Weight
251.39
Test
Specification
APPEARANCE (COLOR)Colorless to Green-Yellow to Yellow
APPEARANCE (FORM)Liquid
PURITY (HPLC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS