Product Name
Benalaxyl PESTANAL®, analytical standard
Product Number
31222
Product Brand
SIAL
CAS Number
71626-11-4
Molecular Weight
325.40
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT75 - 80 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS