Product Name
Fenpropathrin PESTANAL®, analytical standard
Product Number
31223
Product Brand
SIAL
CAS Number
39515-41-8
Molecular Weight
349.42
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT47 - 54 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS