Product Name
Benazolin-ethyl ester PESTANAL®, analytical standard
Product Number
31227
Product Brand
SIAL
CAS Number
25059-80-7
Molecular Weight
271.72
Test
Specification
APPEARANCE (COLOR)White to Faint Yellow to Faint Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT77 - 83 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS