Product Name
Pretilachlor PESTANAL®, analytical standard
Product Number
31251
Product Brand
SIAL
CAS Number
51218-49-6
Molecular Weight
311.85
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow to Brown-Yellow
APPEARANCE (FORM)Liquid
PURITY (HPLC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.5190 - 1.5230
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS