Product Name
Benzo[a]pyrene solution 100 μg/mL in cyclohexane, analytical standard
Product Number
31306
Product Brand
SIAL
CAS Number
50-32-8
Molecular Weight
252.31
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
CONCENTRATION COMP1~ 100 ug/ml
SOLVENTCYCLOHEXANE
PURITY (GC AREA %)≥ 98.0 %
IDENTIFICATION (GC-MS)CORRESPONDS TO REQUIREMENTS
REFRACTIVE INDEX N20/D1.4244 - 1.4284
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS