Product Name
Acrylonitrile analytical standard
Product Number
31564
Product Brand
SIAL
CAS Number
107-13-1
Molecular Weight
53.06
Test
Specification
STABILIZERstabilised
APPEARANCE (COLOR)Colorless to Almost Colorless
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 98.0 %
REFRACTIVE INDEX N20/D1.3890 - 1.3930
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS