Product Name
4-(2-Pyridylazo)resorcinol 96%
Product Number
323209
Product Brand
SIAL
CAS Number
1141-59-9
Molecular Weight
215.21
Test
Specification
APPEARANCE (COLOR)Orange to Brown
APPEARANCE (FORM)Powder
PURITY (HPLC AREA %)≥ 95.5 %
CARBON CONTENT58.6 - 64.2 %
NITROGEN CONTENT18.6 - 20.4 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE