Product Name
Cyclosporin A VETRANAL®, analytical standard
Product Number
32425
Product Brand
SIAL
CAS Number
59865-13-3
Molecular Weight
1202.61
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
IDENTIFICATION (LC-MS)CORRESPONDS TO REQUIREMENTS
WATER≤ 3.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD48 MONTHS