Product Name
Flurprimidol PESTANAL®, analytical standard
Product Number
32523
Product Brand
SIAL
CAS Number
56425-91-3
Molecular Weight
312.29
Test
Specification
APPEARANCE (COLOR)White to Dark Yellow to Dark Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT92 - 98 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD48 MONTHS