Product Name
Quinoclamine PESTANAL®, analytical standard
Product Number
32719
Product Brand
SIAL
CAS Number
2797-51-5
Molecular Weight
207.61
Test
Specification
APPEARANCE (COLOR)Orange to Very Dark Orange to Very Dark Red
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT199 - 205 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS