Product Name
Potassium clavulanate VETRANAL®, analytical standard
Product Number
33454
Product Brand
SIAL
CAS Number
61177-45-5
Molecular Weight
237.25
Test
Specification
PURITY (HPLC AREA %)≥ 98.0 %
WATER≤ 3.0 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
EXPIRATION DATE PERIOD60 MONTHS