Product Name
Pyriftalid PESTANAL®, analytical standard
Product Number
33694
Product Brand
SIAL
CAS Number
135186-78-6
Molecular Weight
318.35
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT161 - 167 C
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS