Product Name
Dimepiperate PESTANAL®, analytical standard
Product Number
33943
Product Brand
SIAL
CAS Number
61432-55-1
Molecular Weight
263.40
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT37 - 42 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS