Product Name
Chlorfenac PESTANAL®, analytical standard
Product Number
34318
Product Brand
SIAL
CAS Number
85-34-7
Molecular Weight
239.48
Test
Specification
APPEARANCE (COLOR)White to Light Yellow
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT157 - 162 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS