Product Name
Cyprodinil PESTANAL®, analytical standard
Product Number
34389
Product Brand
SIAL
CAS Number
121552-61-2
Molecular Weight
225.29
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT74 - 80 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS