Product Name
Flufenamic acid-(benzoic ring-13C6) VETRANAL®, analytical standard
Product Number
35363
Product Brand
SIAL
CAS Number
1325559-30-5
Molecular Weight
287.19
Test
Specification
APPEARANCE (COLOR)White to Green-Yellow to Yellow
APPEARANCE (FORM)Powder or Crystals or Fibers
PURITY (HPLC AREA %)≥ 99.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
RESIDUAL SOLVENTS≤ 1.0 %
REMARKS≥ 98.0% 13C
EXPIRY PERIOD36 MONTHS