Product Name
Metobromuron PESTANAL®, analytical standard
Product Number
36162
Product Brand
SIAL
CAS Number
3060-89-7
Molecular Weight
259.10
Test
Specification
APPEARANCE (COLOR)White to Light Yellow to Light Beige
APPEARANCE (FORM)Powder or Crystals
PURITY (HPLC AREA %)≥ 98.0 %
MELTING POINT95 - 100 C
WATER≤ 1.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS