Product Name
Triclopyr 2-butoxyethylester PESTANAL®, analytical standard
Product Number
36538
Product Brand
SIAL
CAS Number
64700-56-7
Molecular Weight
356.63
Test
Specification
APPEARANCE (COLOR)Colorless to Dark Green-Yellow to Dark Yellow
APPEARANCE (FORM)Liquid or Viscous Liquid
PURITY (HPLC AREA %)≥ 95.0 %
REFRACTIVE INDEX N20/D1.5168 - 1.5208
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD60 MONTHS